Learn More
MTAP Rabbit anti-Human, Unconjugated, Clone: MTAP/3137R, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | MTAP |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MTAP/3137R |
| Conjugate | Unconjugated |
| Description | methylthioadenosine phosphorylase |
| Gene | MTAP |
| Gene Alias | MSAP, c86fus |
| Host Species | Rabbit |
| Immunogen | Recombinant protein corresponding to amino acids 97-196 of human MTAP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.