Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144538
Description
MTB Polyclonal Antibody for Western Blot, ELISA
Probable non-catalytic subunit of the N6-methyltransferase complex, a multiprotein complex that mediates N6-methyladenosine (m6A) methylation at the 5'-[AG]GAC-3' consensus sites of some mRNAs (PubMed:28503769). Associates with MTA, FIP37, VIR and HAKAI to form the m6A writer complex which is essential for adenosine methylation at specific mRNA sequences (PubMed:28503769). N6-methyladenosine (m6A) plays a role in mRNA stability, processing, translation efficiency and editing (PubMed:28503769).
Specifications
MTB | |
Polyclonal | |
Unconjugated | |
MTB | |
EMB1691 | |
Rabbit | |
Affinity chromatography | |
RUO | |
826589 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3.02 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q94AI4 | |
MTB | |
Recombinant Arabidopsis thaliana MTB protein (551-775aa). | |
100 μL | |
Primary | |
Plant | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction