Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTBP Antibody (2C1E11), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250517AF594
Description
MTBP Monoclonal specifically detects MTBP in Human samples. It is validated for Western Blot.Specifications
MTBP | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MTBP | |
Recombinant full-length human MTBP | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Western Blot | |
2C1E11 | |
Western Blot | |
hMTBP, MDM2 (mouse double minute 2)-binding protein, 104kD, Mdm2, transformed 3T3 cell double minute 2, p53 binding protein (mouse) bindingprotein, 104kDa, mdm2-binding protein, MDM2BP | |
Mouse | |
Protein A purified | |
Cancer, DNA Repair | |
27085.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction