Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110909 Supplier Invitrogen™ Supplier No. PA5110909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAKELLKYFT SDGLPIGDLQ PLPIQKGEKT FVLTPELSPG KLQVLPFEKA SVCHYHGIEY CLDDRKALER DGGFSELQSR LIRYETQTTC TR Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 93%.

MTBP inhibits cell migration in vitro and suppresses the invasive behavior of tumor cells. The protein may play a role in MDM2-dependent TP53/p53 homeostasis in unstressed cells. The protein inhibits autoubiquitination of MDM2, thereby enhancing MDM2 stability. This promotes MDM2-mediated ubiquitination of TP53/p53 and its subsequent degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mtbp
Gene Accession No. Q96DY7
Gene Alias AI429604; hdm2; HDMX; hMTBP; MDM2 (mouse double minute 2)-binding protein, 104kD; MDM2 binding protein; Mdm2, transformed 3T3 cell double minute 2, p53 binding protein binding protein, 104kDa; Mdm2, transformed 3T3 cell double minute p53 binding protein; mdm2-binding protein; MDM2BP; MGC5370; MGC71221; mMTBP; Mtbp; RGD1565672
Gene Symbols Mtbp
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human MTBP. Recombinant protein control fragment (Product #RP-93903).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.