Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTCH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113367 Supplier Invitrogen™ Supplier No. PA5113367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTCH1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

MTCH1 is a potential mitochondrial transporter. It may play a role in apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTCH1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.253 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MTCH1
Gene Accession No. Q9NZJ7
Gene Alias 2310034O17Rik; AI255158; AU018396; C77849; cell proliferation-inducing protein 60; CGI-64; mitochondrial carrier 1; mitochondrial carrier homolog 1; mitochondrial carrier homolog 1 (C. elegans); mitochondrial carrier-like protein 1; Mtch1; PIG60; presenilin-associated protein; PSAP; SLC25A49; solute carrier family 25, member 49; UNQ1871/PRO4314
Gene Symbols MTCH1
Host Species Rabbit
Immunogen Recombinant Human Mitochondrial carrier homolog 1 protein (100-250AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23787
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.