Learn More
Invitrogen™ MTCH2 Polyclonal Antibody

Description
A suggested positive control is 293 cell lysate. PA5-20696 can be used with blocking peptide PEP-0811.
Specifications
Specifications
| Antigen | MTCH2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Mtch2 |
| Gene Accession No. | Q791V5, Q9Y6C9 |
| Gene Alias | 2310034D24Rik; 4930539J07Rik; HSPC032; Met-induced mitochondrial protein; MIMP; mitochondrial carrier 2; mitochondrial carrier homolog 2; mitochondrial carrier homolog 2 (C. elegans); Mtch2; SLC25A50; solute carrier family 25, member 50 |
| Gene Symbols | Mtch2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.