Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTCH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558370 Supplier Invitrogen™ Supplier No. PA558370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGYEPLPPTI GRNIFGRQVC QLPGLFSYAQ HIASIDGRRG LFTGLTPRLC SGVLGTVVHG KVLQHYQESD KGEELGP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 88%.

MTCH2 is a member of the mitochondrial carrier protein family that catalyze the exchange of substrates across the inner mitochondrial membrane and is targeted by Bid, a pro-apoptotic Bcl-2 family member, in response to apoptotic signals, suggesting that MTCH2 may play a key role in the mitochondrial apoptosis pathway. Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. The Bcl-2 family of proteins is comprised of critical regulators of apoptosis that can be divided into two classes: those that inhibit apoptosis and those that promote cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTCH2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mtch2
Gene Accession No. Q9Y6C9
Gene Alias 2310034D24Rik; 4930539J07Rik; HSPC032; Met-induced mitochondrial protein; MIMP; mitochondrial carrier 2; mitochondrial carrier homolog 2; mitochondrial carrier homolog 2 (C. elegans); Mtch2; SLC25A50; solute carrier family 25, member 50
Gene Symbols Mtch2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTCH2. Recombinant protein control fragment (Product #RP-97277).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23788
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.