Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTCH2 Polyclonal Antibody

MTCH2 Polyclonal Antibody
Supplier: Thermo Scientific PA578452
Description
MTCH2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| MTCH2 | |
| Polyclonal | |
| Unconjugated | |
| Mtch2 | |
| 2310034D24Rik; 4930539J07Rik; HSPC032; Met-induced mitochondrial protein; MIMP; mitochondrial carrier 2; mitochondrial carrier homolog 2; mitochondrial carrier homolog 2 (C. elegans); Mtch2; SLC25A50; solute carrier family 25, member 50 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23788, 56428 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.49 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q791V5, Q9Y6C9 | |
| Mtch2 | |
| Recombinant protein encompassing a sequence within the center region of human MTCH2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction