Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTCL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561193 Supplier Invitrogen™ Supplier No. PA561193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DRGCGFPVGE HSPHSRVQIG DHSLRLQTAD RGQPHKQVVE NQQLFSAFKA LLEDFRAELR EDERARLRLQ QQYASDKAAW DVEWAVLKCR LEQ Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.

Microtubule-associated factor involved in the late phase of epithelial polarization and microtubule dynamics regulation. Plays a role in the development and maintenance of non-centrosomal microtubule bundles at the lateral membrane in polarized epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTCL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTCL1
Gene Accession No. Q9Y4B5
Gene Alias CCDC165; coiled-coil domain containing 165; coiled-coil domain-containing protein 165; KIAA0802; microtubule crosslinking factor 1; Microtubule cross-linking factor 1; MTCL1; PAR-1-interacting protein; protein SOGA2; SOGA family member 2; SOGA2
Gene Symbols MTCL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTCL1. Recombinant protein control fragment (Product #RP-98267).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.