Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTCO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115081 Supplier Thermo Scientific Supplier No. PA5115081
Add to Cart
Add to Cart

MTCO1 Polyclonal Antibody

MTCO1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MTCO1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene mt-Co1
Gene Accession No. P00395, P00397, P05503
Gene Alias Co1; coi; cox1; coxi; Cytochrome c oxidase polypeptide I; Cytochrome c oxidase subunit 1; cytochrome c oxidase subunit I; mitochondrially encoded cytochrome c oxidase I; mtco1; mt-co1
Gene Symbols mt-Co1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MT-CO1(Accession P00395), corresponding to amino acid residues H413-T463.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17708, 26195, 4512
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.