Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTCO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501735966 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5966 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5966 Supplier Proteintech Group Inc Supplier No. 550701AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody is specific to COX2.

Cytochrome c oxidase is the fourth complex in the respiratory chain and is responsible for catalyzing the conversion of O2 to H2O. Subunit 2 of the cytochrome c oxidase complex combines with two other subunits (1 and 3) to form a core protein structure that performs many functions of the enzyme. The metallic copper center of this subunit transfers electrons to the heme center of subunit 1, which results in the movement of electrons from cytochrome c to the heme A3 and copper B metallic center of complex IV. Defects in subunit 2 of cytochrome c oxidase can result in COX deficiency, which causes a wide range of symptoms from local myopathy to multiple system pathologies that begin between infancy and adulthood. Abnormalities in this subunit are also associated with tumor development.

Specifications

Antigen MTCO2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COX2
Gene Accession No. P00403, P00405
Gene Alias COII, COX2, COXII, cytochrome c oxidase II, Cytochrome c oxidase subunit 2, MT CO2, MTCO2
Gene Symbols COX2
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17709, 26198, 4513
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.