Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTCP1 Mouse anti-Human, Clone: BM-3, Novus Biologicals™

Mouse Monoclonal Antibody
$231.50 - $499.50
Specifications
Antigen | MTCP1 |
---|---|
Clone | BM-3 |
Dilution | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Description
MTCP1 Monoclonal antibody specifically detects MTCP1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
MTCP1 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
Monoclonal | |
Purified | |
RUO | |
Human | |
mature T-cell proliferation 1, P13MTCP1 | |
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with nuclei from Pokeweed mitogen stimulated with peripheral blood lymphocytes and mouse myeloma cells. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
BM-3 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
PBS | |
4515 | |
IgG1 | |
Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title