Learn More
Invitrogen™ MTCP1 Polyclonal Antibody

Description
MTCP1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | MTCP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | MTCP1 |
| Gene Accession No. | P56278, Q60945 |
| Gene Alias | C6.1b; C61B; Cmc4; mature T cell proliferation 1; mature T-cell proliferation 1; mature T-cell proliferation 1 isoform p13; mature T-cell proliferation 1 neighbor protein; mature T-cell proliferation-1 type A; mature T-cell proliferation-1 type B1; MTCP1; MTCP-1 type A; MTCP-1 type B1; p13MTCP1; p8MTCP1; Protein p13 MTCP-1; protein p8 MTCP-1 |
| Gene Symbols | MTCP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.