Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTERF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567057
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567057 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567057 Supplier Invitrogen™ Supplier No. PA567057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSLGQTSISKG LNYLTIMAPG NLWHMRNNFL FGSRCWMTRF SAENIFKSVS FRLFGVKCHN TDSEPLKNED LLKNLLTMGV DIDMARKRQP GVFHRMITNE QDLKMFLLSK GASKEVIASI ISRYPRAITR TPENLSKRWD LWRKIVTSD Highest antigen sequence identity to the following orthologs - mouse 61%, rat 61%.

Transcription termination factor. Binds to a 28 bp region within the tRNA(Leu(uur)) gene at a position immediately adjacent to and downstream of the 16S rRNA gene; this region comprises a tridecamer sequence critical for directing accurate termination. Binds DNA along the major grove and promotes DNA bending and partial unwinding. Promotes base flipping. Transcription termination activity appears to be polarized with highest specificity for transcripts initiated on the light strand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTERF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTERF1
Gene Accession No. Q99551
Gene Alias 4931431L11Rik; 9230106K09Rik; mitochondrial transcription termination factor; mitochondrial transcription termination factor 1; Mitochondrial transcription termination factor 1a; mitochondrial transcription termination factor 1b; mTERF; Mterf1; mTERF1a; mTERF1b; transcription termination factor 1, mitochondrial; Transcription termination factor 1a, mitochondrial; transcription termination factor, mitochondrial
Gene Symbols MTERF1
Host Species Rabbit
Immunogen Recombinant Human MTERF. Recombinant protein control fragment (Product #RP-105478).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.