Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTERFD1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281740AF750
This item is not returnable.
View return policy
Description
MTERFD1 Polyclonal antibody specifically detects MTERFD1 in Human, Mouse samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
MTERFD1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 750 | |
CGI-12, FLJ10939, Mitochondrial transcription termination factor 3, MTERF domain containing 1, mTERF domain-containing protein 1, mitochondrial, mTERF3 | |
MTERFD1 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MTERFD1 . The immunogen is located within the last 50 amino acids of MTERFD1. Amino Acid Squence: LGRAQYDPAKPNYISLD | |
0.1 mL | |
Autophagy, Cell Biology, Endocrinology | |
51001 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction