Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTERFD1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281740PCP
Description
MTERFD1 Polyclonal antibody specifically detects MTERFD1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MTERFD1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| CGI-12, FLJ10939, Mitochondrial transcription termination factor 3, MTERF domain containing 1, mTERF domain-containing protein 1, mitochondrial, mTERF3 | |
| MTERFD1 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MTERFD1 . The immunogen is located within the last 50 amino acids of MTERFD1. Amino Acid Squence: LGRAQYDPAKPNYISLD | |
| 0.1 mL | |
| Autophagy, Cell Biology, Endocrinology | |
| 51001 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction