Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTERFD1 Monoclonal Antibody (A8-A10)
GREENER_CHOICE

Catalog No. PIMA532845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532845 Supplier Invitrogen™ Supplier No. MA532845
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTERFD1 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD1 is composed of three leucine zippers that form a three-stranded coiled-coil that binds to DNA. It binds promoter DNA and acts as a mitochondrial transcription regulator. It has been suggested that only the phosphorylated form of MTERFD1 has transcription termination activity. A closely related mTERF family member, MTERFD3, is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTERFD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone A8-A10
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4
Gene MTERF3
Gene Accession No. Q8R3J4, Q96E29
Gene Alias 2410017I18Rik; Cgi12; CGI-12; Mitochondrial transcription termination factor 3; MTERF domain containing 1; mTERF domain-containing protein 1, mitochondrial; Mterf3; MTERFD1; transcription termination factor 3, mitochondrial
Gene Symbols MTERF3
Host Species Mouse
Immunogen Recombinant protein with Human MTERFD1 aa 29-417.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51001, 66410
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.