Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTERFD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534530
Description
A suggested positive control is NIH-3T3 cell lysate. PA5-34530 can be used with blocking peptide PEP-1573.
Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD1 is composed of three leucine zippers that form a three-stranded coiled-coil that binds to DNA. It binds promoter DNA and acts as a mitochondrial transcription regulator. It has been suggested that only the phosphorylated form of MTERFD1 has transcription termination activity. A closely related mTERF family member, MTERFD3, is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription.
Specifications
MTERFD1 | |
Polyclonal | |
Unconjugated | |
MTERF3 | |
2410017I18Rik; Cgi12; CGI-12; Mitochondrial transcription termination factor 3; MTERF domain containing 1; mTERF domain-containing protein 1, mitochondrial; Mterf3; MTERFD1; transcription termination factor 3, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51001, 66410 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8R3J4, Q96E29 | |
MTERF3 | |
A 17 amino acid peptide near the carboxy terminus of human MTERFD1. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction