Learn More
Invitrogen™ MTERFD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597246
Description
Immunogen sequence: SSQSTSSSSQ ENNSAQSSLL PSMNEQSQKT QNISSFDSEL FLEELDELPP LSPMQPISEE EAIQIIADPP LPPASFTLRD YVDHSETLQK LVLLGVDLSK IEKHPEAANL LLRLDFEKDI KQMLLFLKDV GIEDNQLGAF LTKNHAIFSE DLENLKTRVA YLHSKNFSKA DVAQMVRKAP FLLNFSVERL DNRLGFFQKE LELSVKKTRD LVVRLPRLLT GSLEPVKENM KV; Positive Samples: HepG2, 293T, SH-SY5Y, NCI-H460, SKOV3, BT-474; Cellular Location: Mitochondrion.
Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD1 is composed of three leucine zippers that form a three-stranded coiled-coil that binds to DNA. It binds promoter DNA and acts as a mitochondrial transcription regulator. It has been suggested that only the phosphorylated form of MTERFD1 has transcription termination activity. A closely related mTERF family member, MTERFD3, is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription.
Specifications
MTERFD1 | |
Polyclonal | |
Unconjugated | |
MTERF3 | |
2410017I18Rik; Cgi12; CGI-12; Mitochondrial transcription termination factor 3; MTERF domain containing 1; mTERF domain-containing protein 1, mitochondrial; Mterf3; MTERFD1; transcription termination factor 3, mitochondrial | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51001, 66410 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R3J4, Q96E29 | |
MTERF3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 69-300 of human MTERFD1 (NP_0570263). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.