Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTERFD1 Polyclonal Antibody
MTERFD1 Polyclonal Antibody
Supplier: Thermo Scientific PA598630
Description
MTERFD1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
MTERFD1 | |
Polyclonal | |
Unconjugated | |
MTERF3 | |
2410017I18Rik; Cgi12; CGI-12; Mitochondrial transcription termination factor 3; MTERF domain containing 1; mTERF domain-containing protein 1, mitochondrial; Mterf3; MTERFD1; transcription termination factor 3, mitochondrial | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51001 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96E29 | |
MTERF3 | |
Recombinant Human Transcription termination factor 3, mitochondrial protein (69-300AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction