Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTERFD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113083
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113083 Supplier Invitrogen™ Supplier No. PA5113083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTERFD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD2 is targeted to the mitochondria and is ubiquitously expressed, with highest expression levels in fore- and midbrain, diencephalon, spinal cord, tongue, lung liver and kidney. MTERFD2 has been suggested to play a role in organ differentiation during embryogenesis. A closely related mTERF family member, MTERFD3, is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTERFD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MTERF4
Gene Accession No. Q7Z6M4, Q8BVN4
Gene Alias 1810059A23Rik; 4933412H03Rik; HSPC096; Mitochondrial transcription termination factor 4; MTERF domain containing 2; mTERF domain-containing protein 2; mTERF domain-containing protein 2 processed; MTERF4; Mterfd2; Transcription termination factor 4, mitochondrial
Gene Symbols MTERF4
Host Species Rabbit
Immunogen Recombinant Human Transcription termination factor 4, mitochondrial protein (45-87AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130916, 69821
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.