Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTERFD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534531
Description
A suggested positive control is human small intestine tissue lysate. PA5-34531 can be used with blocking peptide PEP-1574.
Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD2 is targeted to the mitochondria and is ubiquitously expressed, with highest expression levels in fore- and midbrain, diencephalon, spinal cord, tongue, lung liver and kidney. MTERFD2 has been suggested to play a role in organ differentiation during embryogenesis. A closely related mTERF family member, MTERFD3, is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription.
Specifications
| MTERFD2 | |
| Polyclonal | |
| Unconjugated | |
| MTERF4 | |
| 1810059A23Rik; 4933412H03Rik; HSPC096; Mitochondrial transcription termination factor 4; MTERF domain containing 2; mTERF domain-containing protein 2; mTERF domain-containing protein 2 processed; MTERF4; Mterfd2; Transcription termination factor 4, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 130916, 363289, 69821 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q4G078, Q7Z6M4, Q8BVN4 | |
| MTERF4 | |
| A 17 amino acid peptide near the carboxy terminus of human MTERFD2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction