Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTERFD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534532
Description
A suggested positive control is human testis tissue lysate. PA5-34532 can be used with blocking peptide PEP-1575.
Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD3 was initially identified through differential display analysis and is expressed primarily in heart, liver, pancreas and skeletal muscle. MTERFD3 is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription, and may be a serum-inhibitory factor.
Specifications
MTERFD3 | |
Polyclonal | |
Unconjugated | |
MTERF2 | |
1700007D05Rik; mitochondrial transcription termination factor 2; Mitochondrial transcription termination factor-like protein; MTERF domain containing 3; mTERF domain-containing protein 3, mitochondrial; MTERF2; Mterfd3; mTERFL; mTERF-like; RGD1311836; Transcription termination factor 2, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
74238, 80298 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q49AM1, Q8BKY8 | |
MTERF2 | |
An 18 amino acid peptide near the amino terminus of human MTERFD3. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction