Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTERFD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551952 Supplier Invitrogen™ Supplier No. PA551952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TRTVEKLYKC SVDIRKIRRL KGWVLLEDET YVEEIANILQ ELGADETAVA SILERCPEAI VCSPTAVNTQ RKLWQLVCKN EEELIKLIEQ FPESFFTIKD QENQKLNVQF FQELGLKNVV ISRLLTAAPN VFHNPVEKNK QMVR Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.

Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD3 was initially identified through differential display analysis and is expressed primarily in heart, liver, pancreas and skeletal muscle. MTERFD3 is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription, and may be a serum-inhibitory factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTERFD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTERF2
Gene Accession No. Q49AM1
Gene Alias 1700007D05Rik; mitochondrial transcription termination factor 2; Mitochondrial transcription termination factor-like protein; MTERF domain containing 3; mTERF domain-containing protein 3, mitochondrial; MTERF2; Mterfd3; mTERFL; mTERF-like; RGD1311836; Transcription termination factor 2, mitochondrial
Gene Symbols MTERF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTERFD3. Recombinant protein control fragment (Product #RP-102232).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80298
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.