Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTF1 Antibody (OTI2F3), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272796UV
Description
MTF1 Monoclonal specifically detects MTF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MTF1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MTF1 | |
Recombinant protein fragment corresponding to amino acids 1-272 of human MTF1 (NP_005946) produced in E.coli. | |
0.1 mL | |
4520.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2F3 | |
Western Blot, Immunohistochemistry | |
metal regulatory transcription factor 1, metal-regulatory transcription factor 1, metal-responsive transcription factor 1, MGC23036, MRE-binding transcription factor, MRE-binding transcription factor-1, MTF-1, Transcription factor MTF-1, zinc regulatory factor, ZRF | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction