Learn More
MTF1 Mouse anti-Human, Clone: OTI2F3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805480

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a transcription factor that induces expression of metallothioneins and other genes involved in metal homeostasis in response to heavy metals such as cadmium, zinc, copper, and silver. The protein is a nucleocytoplasmic shuttling protein that accumulates in the nucleus upon heavy metal exposure and binds to promoters containing a metal-responsive element (MRE).Specifications
MTF1 | |
Monoclonal | |
Unconjugated | |
MTF1 | |
MTF-1, ZRF | |
Mouse | |
Affinity Chromatography | |
RUO | |
4520 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2F3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14872 | |
MTF1 | |
Human recombit protein fragment corresponding to amino acids 1-272 of human MTF1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.