Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540475 Supplier Invitrogen™ Supplier No. PA540475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QSSLVMGEQN LQWILNGATS SPQNQEQIQQ ASKVEKVFFT TAVPVASSPG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

This gene encodes a transcription factor that induces expression of metallothioneins and other genes involved in metal homeostasis in response to heavy metals such as cadmium, zinc, copper, and silver. The protein is a nucleocytoplasmic shuttling protein that accumulates in the nucleus upon heavy metal exposure and binds to promoters containing a metal-responsive element (MRE).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTF1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MTF1
Gene Accession No. Q14872
Gene Alias metal regulatory transcription factor 1; metal response element binding transcription factor 1; metalloregulatory transcription factor; metal-regulatory transcription factor 1; metal-responsive transcription factor 1; MRE-binding transcription factor; MRE-binding transcription factor-1; Mtf1; MTF-1; Thyls; Transcription factor MTF-1; zinc regulatory factor; ZRF
Gene Symbols MTF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human MTF1 (aa 562-611).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4520
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.