Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTFMT Antibody (OTI2A2), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272803C
Description
MTFMT Monoclonal specifically detects MTFMT in Human samples. It is validated for Western Blot.Specifications
MTFMT | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MTFMT | |
Recombinant protein fragment corresponding to amino acids 175-389 of human MTFMT (NP_640335) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2A2 | |
Western Blot | |
EC 2.1.2.9, FMT, FMT1methionyl-tRNA formyltransferase, mitochondrial, mitochondrial methionyl-tRNA formyltransferase, MtFMT | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
123263.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction