Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTFMT Monoclonal Antibody (OTI1D4), TrueMAB™, OriGene
SDP

Catalog No. 502540847
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0847 30 μL
50-167-4412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0847 Supplier OriGene Supplier No. TA503548S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTFMT Monoclonal antibody specifically detects MTFMT in Human samples. It is validated for Western Blot

Specifications

Antigen MTFMT
Applications Western Blot
Classification Monoclonal
Clone OTI1D4
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MTFMT
Gene Accession No. Q96DP5
Gene Alias 2310020P08Rik; COXPD15; Fmt; FMT1; LOW QUALITY PROTEIN: methionyl-tRNA formyltransferase, mitochondrial; methionyl-tRNA formyltransferase, mitochondrial; mitochondrial methionyl-tRNA formyltransferase; mitochondrial methionyl-tRNA transformylase; MtFMT; RGD1309462
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 175-389 of human MTFMT produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123263
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.