Learn More
MTFMT Mouse anti-Human, Clone: OTI1C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503547

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this nuclear gene localizes to the mitochondrion, where it catalyzes the formylation of methionyl-tRNA.Specifications
MTFMT | |
Monoclonal | |
Unconjugated | |
MTFMT | |
COXPD15, FMT1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
123263 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96DP5 | |
MTFMT | |
Human recombit protein fragment corresponding to amino acids 175-389 of human MTFMT produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.