Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTFMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | MTFMT |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MTFMT Polyclonal antibody specifically detects MTFMT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
MTFMT | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.1.2.9, FMT, FMT1methionyl-tRNA formyltransferase, mitochondrial, mitochondrial methionyl-tRNA formyltransferase, MtFMT | |
Recombinant fusion protein containing a sequence corresponding to amino acids 180-389 of human MTFMT (NP_640335.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, 50% glycerol, pH7.3 | |
123263 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title