Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTFP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567454 Supplier Invitrogen™ Supplier No. PA567454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CFMSTSYSCQG MWTPGSLVSK DPGTWVGLSW TEA Highest antigen sequence identity to the following orthologs - mouse 35%, rat 35%.

MTP18 (mitochondrial 18 kDa protein), also known as HSPC242, is a 166 amino acid protein essential for cell viability. MTP18 may also have a role in maintaining the integrity of the mitochondrial network. Localized to the mitochonrion, expression of MTP18 is regulated by the PI 3-kinase pathway. Loss of function of MTP18 results in the release of cytochrome c, which subsequently activates the caspase cascade and leads to apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTFP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mtfp1
Gene Accession No. Q9UDX5
Gene Alias 1700020C11Rik; 2610507A21Rik; AV005788; HSPC242; Mitochondrial 18 kDa protein; mitochondrial fission process 1; mitochondrial fission process protein 1; mitochondrial fission protein MTP18; mitochondrial protein 18 kDa; MTFP1; Mtp18; My022
Gene Symbols Mtfp1
Host Species Rabbit
Immunogen Recombinant Human MTFP1. Recombinant protein control fragment (Product #RP-108112).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51537
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.