Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTFR1 Polyclonal Antibody
MTFR1 Polyclonal Antibody
Supplier: Thermo Scientific PA570577
Description
MTFR1 Polyclonal Antibody for Western Blot

Specifications
MTFR1 | |
Polyclonal | |
Unconjugated | |
Mtfr1 | |
1300002C08Rik; 1700080D04Rik; 2810026O10Rik; 4930579E05Rik; chondrocyte protein with a poly-proline region; CHPPR; FAM54A2; Kiaa0009; Mitochondrial fission regulator 1; Mtfr1; RGD1304936 | |
Rabbit | |
Affinity chromatography | |
RUO | |
9650 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15390 | |
Mtfr1 | |
synthetic peptide directed towards the middle region of human MTFR1 (aa 199-248). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction