Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTFR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584278 Supplier Invitrogen™ Supplier No. PA584278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLHQSTSAVD LIKERREKRA NAGKTLVKNN PKKPEMPNML EILKEMNSVK LRSVKRSEQD V Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 75%.

This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTFR1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mtfr1
Gene Accession No. Q15390
Gene Alias 1300002C08Rik; 1700080D04Rik; 2810026O10Rik; 4930579E05Rik; chondrocyte protein with a poly-proline region; CHPPR; FAM54A2; Kiaa0009; Mitochondrial fission regulator 1; Mtfr1; RGD1304936
Gene Symbols Mtfr1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTFR1. Recombinant protein control fragment (Product #RP-105034).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9650
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.