Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558128
Description
Immunogen sequence: KLVDCIIEVH DARIPLSGRN PLFQETLGLK PHLLVLNKMD LADLTEQQKI MQHLEGEGLK NVIFTNCVKD ENVKQIIPMV TELIGRSHRY H Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 87%.
Mitochondrial GTPase. May be involved in assembly of the large ribosomal subunit.
Specifications
MTG1 | |
Polyclonal | |
Unconjugated | |
MTG1 | |
Gm169; GTP; GTP binding protein 7 (putative); GTP-binding protein 7; GTP-binding protein 7 (putative); GTPBP7; Mitochondrial GTPase 1; mitochondrial GTPase 1 homolog; mitochondrial GTPase 1 homolog (S. cerevisiae); mitochondrial GTPase 1-like; mitochondrial ribosome associated GTPase 1; mitochondrial ribosome-associated GTPase 1; Mtg1; RP11-108K14.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
92170 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BT17 | |
MTG1 | |
Recombinant protein corresponding to Human MTG1. Recombinant protein control fragment (Product #RP-97110). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction