Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118091 Supplier Invitrogen™ Supplier No. PA5118091
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MTG1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Mitochondrial GTPase. May be involved in assembly of the large ribosomal subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTG1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene MTG1
Gene Accession No. Q9BT17
Gene Alias Gm169; GTP; GTP binding protein 7 (putative); GTP-binding protein 7; GTP-binding protein 7 (putative); GTPBP7; Mitochondrial GTPase 1; mitochondrial GTPase 1 homolog; mitochondrial GTPase 1 homolog (S. cerevisiae); mitochondrial GTPase 1-like; mitochondrial ribosome associated GTPase 1; mitochondrial ribosome-associated GTPase 1; Mtg1; RP11-108K14.2
Gene Symbols MTG1
Host Species Rabbit
Immunogen E. coli-derived Human MTG1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92170
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.