Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTG16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566489 Supplier Thermo Scientific Supplier No. PA566489
Add to Cart
Add to Cart

MTG16 Polyclonal Antibody

MTG16 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen MTG16
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CBFA2T3
Gene Accession No. O75081
Gene Alias A630044F12Rik; AI465270; AW229127; CBFA2/RUNX1 partner transcriptional co-repressor 3; CBFA2/RUNX1 translocation partner 3; CBFA2T3; Cbfa2t3h; core-binding factor, runt domain, alpha subunit 2, translocated to, 3; core-binding factor, runt domain, alpha subunit 2, translocated to, 3 (human); core-binding factor, runt domain, alpha subunit 2; translocated to, 3; eight twenty one protein 2; ETO/MTG8-related protein ETO-2; Eto2; ETO-2; hMTG16; MTG16; MTG16HEL; MTG8-related gene 2; MTG8-related protein 2; MTGR2; myeloid translocation gene 8 and 16b; myeloid translocation gene on chromosome 16 protein; myeloid translocation gene-related protein 2; protein CBFA2T3; Protein ETO-2; RUNX1T3; translocated to, 3; zinc finger MYND domain-containing protein 4; ZMYND4
Gene Symbols CBFA2T3
Host Species Rabbit
Immunogen Recombinant Human MTG16. Recombinant protein control fragment (Product #RP-107162).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 863
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.