Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTGR1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197748FR
Description
MTGR1 Polyclonal antibody specifically detects MTGR1 in Human samples. It is validated for Western BlotSpecifications
MTGR1 | |
Polyclonal | |
Western Blot | |
core-binding factor, runt domain, alpha subunit 2; translocated to, 2, EHTDKFZp313F2116, ETO homolog on chromosome 20, ETO homologous on chromosome 20, MTG8-like protein, MTG8-related protein 1, MTGR1p85, myeloid translocation gene-related protein 1, Myeloid translocation-related protein 1, protein CBFA2T2, ZMYND3 | |
The immunogen recognized by this antibody maps to a region between residue 554 and 604 of human Core-Binding Factor, Runt Domain, Alpha Subunit 2; translocated to, 2 using the numbering given in entry NP_005084.1 (GeneID 9139). | |
0.1 mL | |
Cancer, Tumor Suppressors | |
9139 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction