Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTGR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674141
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-141 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-141 Supplier Invitrogen™ Supplier No. PA112415
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

MTGR1 Polyclonal Antibody for Western Blot

Myeloid tumor related gene (MTGR1) is found deleted in acute myeloid leukemia. Overexpression of MTGR1 can stimulate AML1-MTG8 to induce GCSF- dependent proliferation of L-G cells and to interfere with AML1- dependent transcription. AML1/MTG8 functions as a complex with MTGR1 and that the complex might be important in promoting leukemogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTGR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene CBFA2T2
Gene Accession No. O43439
Gene Alias A430091M07; C330013D05Rik; CBFA2/RUNX1 partner transcriptional co-repressor 2; CBFA2/RUNX1 translocation partner 2; CBFA2T2; CBFA2T2 identified gene homolog; Cbfa2t2h; core-binding factor runt domain alpha subunit 2 translocated to 2 homolog; core-binding factor, runt domain, alpha subunit 2, translocated to, 2 (human); core-binding factor, runt domain, alpha subunit 2, translocated to, 2 homolog; core-binding factor, runt domain, alpha subunit 2; translocated to, 2; DKFZp313F2116; EHT; ETO homolog on chromosome 20; ETO homologous on chromosome 20; MTG8-like protein; MTG8-related protein 1; MTGR1; myeloid translocation gene-related protein 1; myeloid translocation-related protein 1; p85; Protein CBFA2T2; RP5-1137F22.1; translocated to, 2; ZMYND3; ZMYND3 CBFA2T2
Gene Symbols CBFA2T2
Host Species Sheep
Immunogen C-terminal portion of recombinant MTGR1.
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9139
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.