Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTGR1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988449
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8449 Supplier Bioss Supplier No. BS8304R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myeloid tumor related gene (MTGR1) is found deleted in acute myeloid leukemia. Overexpression of MTGR1 can stimulate AML1-MTG8 to induce GCSF- dependent proliferation of L-G cells and to interfere with AML1- dependent transcription. AML1/MTG8 functions as a complex with MTGR1 and that the complex might be important in promoting leukemogenesis.

Specifications

Antigen MTGR1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CBFA2T2
Gene Accession No. O43439
Gene Alias A430091M07; C330013D05Rik; CBFA2/RUNX1 translocation partner 2; CBFA2T2; CBFA2T2 identified gene homolog; Cbfa2t2h; core-binding factor runt domain alpha subunit 2 translocated to 2 homolog; core-binding factor, runt domain, alpha subunit 2, translocated to, 2 (human); core-binding factor, runt domain, alpha subunit 2, translocated to, 2 homolog; core-binding factor, runt domain, alpha subunit 2; translocated to, 2; DKFZp313F2116; EHT; ETO homolog on chromosome 20; ETO homologous on chromosome 20; MTG8-like protein; MTG8-related protein 1; Mtgr1; myeloid translocation gene-related protein 1; Myeloid translocation-related protein 1; p85; Protein CBFA2T2; RP5-1137F22.1; translocated to, 2; ZMYND3; ZMYND3 CBFA2T2
Gene Symbols CBFA2T2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MTGR1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9139
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.