Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTH1 Antibody (AT3B3) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$340.50 - $499.50

Specifications

Antigen MTH1
Clone AT3B3
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP26116050
SDP
View Documents
Novus Biologicals
NBP26116050UL
50 μL
Each for $340.50
Only null left
Add to Cart
 
NB261160100
SDP
View Documents
Novus Biologicals
NBP261160100UL
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

MTH1 Monoclonal specifically detects MTH1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MTH1
1 mg/mL
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Human
8-oxo-7,8-dihydrodeoxyguanosine triphosphatase, EC 3.6.1.-, MTH1Nucleoside diphosphate-linked moiety X motif 1, mutT human homolog 1,8-oxo-dGTPase, nucleoside diphosphate-linked moiety X-type motif 17,8-dihydro-8-oxoguanine triphosphatase, nudix (nucleoside diphosphate linked moiety X)-type motif 1,8-oxo-7,8-dihydroguanosine triphosphatase, Nudix motif 1
NUDT1
IgG1 κ
Protein A or G purified
AT3B3
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500
Monoclonal
Purified
Base Excision Repair, Cancer, Chromatin Research, DNA Repair, Modulation of DNA Pools, Stem Cell Markers
PBS (pH7.4) w/ 10% glycerol with 0.02% Sodium Azide
4521
Recombinant human NUDT1 (1-156aa) purified from E. coli
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.