Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTH1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $560.50
Specifications
Antigen | MTH1 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500, Simple Western 1:50, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200, Immunoblotting reported in scientific literature (PMID 21076467), Knockout Validated, Single Cell Western 100 ug/ml, KnockDown Validated |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100109SS
![]() |
Novus Biologicals
NB100109SS |
0.05 mL |
Each for $243.50
|
|
|||||
NB100109
![]() |
Novus Biologicals
NB100109 |
0.2 mL |
Each for $560.50
|
|
|||||
Description
MTH1 Polyclonal specifically detects MTH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Knockout Validated, Single Cell Western, Knockdown Validated.Specifications
MTH1 | |
Western Blot 1:500, Simple Western 1:50, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200, Immunoblotting reported in scientific literature (PMID 21076467), Knockout Validated, Single Cell Western 100 ug/ml, KnockDown Validated | |
Polyclonal | |
Rabbit | |
Base Excision Repair, Cancer, Chromatin Research, DNA Repair, Modulation of DNA Pools, Stem Cell Markers | |
PBS with 0.02% Sodium Azide | |
8-oxo-7,8-dihydrodeoxyguanosine triphosphatase, EC 3.6.1.-, MTH1Nucleoside diphosphate-linked moiety X motif 1, mutT human homolog 1,8-oxo-dGTPase, nucleoside diphosphate-linked moiety X-type motif 17,8-dihydro-8-oxoguanine triphosphatase, nudix (nucleoside diphosphate linked moiety X)-type motif 1,8-oxo-7,8-dihydroguanosine triphosphatase, Nudix motif 1 | |
NUDT1 | |
IgG | |
Affinity Purified | |
18 kDa |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
P36639 | |
4521 | |
A C-terminal peptide derived from human MTH1, conjugated to KLH. [UniProt# P36639] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title