Learn More
Invitrogen™ MTH1 Polyclonal Antibody

Description
Immunogen sequence: MSGISPQQMG EPEGSWSGKN PGTMGASRLY TLVLVLQPQR VLLGMKKRGF GAGRWNGFGG KVQEGETIED GARRELQEES GLTVDALHKV GQIVFEFVGE PELMDVHVFC TDSIQGTPVE SDEMRPCWFQ LDQIPFKDMW PDDSYWFPLL LQKKKFHGYF KFQGQDTILD YTLREVDTV; Positive Samples: SW480, K562, 22RV1, THP-1, HepG2, SKOV3, 293T; Cellular Location: Cytoplasm, Mitochondrion matrix, Nucleus.
Specifications
Specifications
| Antigen | MTH1 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.94 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NUDT1 |
| Gene Accession No. | P36639 |
| Gene Alias | 2-hydroxy-dATP diphosphatase; 7,8-dihydro-8-oxoguanine triphosphatase; 8-oxo-7,8-dihydro-2'-dGTPase; 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase; 8-oxo-7,8-dihydroguanosine triphosphatase; 8-oxo-dGTPase; Mth1; mutT human homolog (8-oxo-dGTPase); mutT human homolog 1; nucleoside diphosphate-linked moiety X motif 1; nucleoside diphosphate-linked moiety X-type motif 1; nudix (nucleoside diphosphate linked moiety X)-type motif 1; nudix hydrolase 1; nudix motif 1; nudix-type motif 1; NUDT1 |
| Gene Symbols | NUDT1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.