Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTHFD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542825 Supplier Invitrogen™ Supplier No. PA542825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RTTTESEVMK YITSLNEDST VHGFLVQLPL DSENSINTEE VINAIAPEKD Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 100%; Zebrafish: 85%.

This gene encodes a protein that possesses three distinct enzymatic activities, 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methenyltetrahydrofolate cyclohydrolase and 10-formyltetrahydrofolate synthetase. Each of these activities catalyzes one of three sequential reactions in the interconversion of 1-carbon derivatives of tetrahydrofolate, which are substrates for methionine, thymidylate, and de novo purine syntheses. The trifunctional enzymatic activities are conferred by two major domains, an aminoterminal portion containing the dehydrogenase and cyclohydrolase activities and a larger synthetase domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTHFD1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MTHFD1
Gene Accession No. P11586
Gene Alias 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methylenetetrahydrofolate cyclohydrolase, 10-formyltetrahydrofolate synthetase; C1-tetrahydrofolate synthase; C-1-tetrahydrofolate synthase, cytoplasmic; C-1-tetrahydrofolate synthase, cytoplasmic, N-terminally processed; C1-THF synthase; cytoplasmic C-1-tetrahydrofolate synthase; Dcs; E430024A07Rik; Formyltetrahydrofolate synthetase; LOW QUALITY PROTEIN: C-1-tetrahydrofolate synthase, cytoplasmic; methenyltetrahydrofolate; methenyltetrahydrofolate cyclohydrolase; Methylenetetrahydrofolate dehydrogenase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthetase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent), methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthase; methylenetetrahydrofolate dehydrogenase 1; methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1; MTHFC; MTHFD; MTHFD1; MTHFD1 protein; formyltetrahydrofolate synthetase; methenyltetrahydrofolate cyclohydrolase,
Gene Symbols MTHFD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MTHFD1 (aa 74-123).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4522
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.