Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTHFD1L Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP178279
Description
MTHFD1L Polyclonal specifically detects MTHFD1L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
MTHFD1L | |
Polyclonal | |
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
10-formyl-THF synthetase, dJ292B18.2, DKFZp586G1517, EC 6.3.4.3, FLJ21145, Formyltetrahydrofolate synthetase, formyltetrahydrofolate synthetase domain containing 1, FTHFSDC1, methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like, mitochondrial C1-tetrahydrofolate synthase, mitochondrial C1-tetrahydrofolate synthetase, monofunctional C1-tetrahydrofolate synthase, mitochondrial, MTC1THFS | |
Goat | |
Affinity Purified | |
RUO | |
25902 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
MTHFD1L | |
Peptide with sequence C-DTNDRFLRKITIGQ corresponding to internal region according to NP_056255.2. | |
0.1 mg | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction