Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTHFD1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576315
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576315 Supplier Invitrogen™ Supplier No. PA576315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

One-carbon substituted forms of tetrahydrofolate (THF) are involved in the de novo synthesis of purines and thymidylate and support cellular methylation reactions through the regeneration of methionine from homocysteine. MTHFD1L is an enzyme involved in THF synthesis in mitochondria (Christensen et al., 2005 [PubMed 15611115]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTHFD1L
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MTHFD1L
Gene Accession No. Q3V3R1, Q6UB35
Gene Alias 10-formyl-THF synthetase; 2410004L15Rik; AI647056; dJ292B18.2; Formyltetrahydrofolate synthetase; formyltetrahydrofolate synthetase domain containing 1; FTHFSDC1; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like; monofunctional C1-tetrahydrofolate synthase, mitochondrial; MTC1THFS; MTHFD1L
Gene Symbols MTHFD1L
Host Species Rabbit
Immunogen Recombinant protein of human MTHFD1L.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25902, 270685, 361472
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.