Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTHFD2 Monoclonal Antibody (1985CT220.118.31)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537767
Description
MTHFD2 Monoclonal Antibody for Western Blot
This gene encodes a nuclear-encoded mitochondrial bifunctional enzyme with methylenetetrahydrofolate dehydrogenase and methenyltetrahydrofolate cyclohydrolase activities. The enzyme functions as a homodimer and is unique in its absolute requirement for magnesium and inorganic phosphate. Formation of the enzyme-magnesium complex allows binding of NAD. Alternative splicing results in two different transcripts, one protein-coding and the other not protein-coding. This gene has a pseudogene on chromosome 7.
Specifications
MTHFD2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P13995, P18155 | |
MTHFD2 | |
KLH conjugated synthetic peptide between 266-299 amino acids from the C-terminal region of human MTHFD2. | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Western Blot | |
1985CT220.118.31 | |
Unconjugated | |
MTHFD2 | |
AW558851; Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase, mitochondrial; Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NAD+ dependent), methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase 2; Mthfd2; NAD-dependent methylene tetrahydrofolate dehydrogenase cyclohydrolase; NAD-dependent methylenetetrahydrofolate dehydrogenase; NMDMC | |
Mouse | |
Protein G | |
RUO | |
10797, 17768 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction