Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTHFD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576502
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a nuclear-encoded mitochondrial bifunctional enzyme with methylenetetrahydrofolate dehydrogenase and methenyltetrahydrofolate cyclohydrolase activities. The enzyme functions as a homodimer and is unique in its absolute requirement for magnesium and inorganic phosphate. Formation of the enzyme-magnesium complex allows binding of NAD. Alternative splicing results in two different transcripts, one protein-coding and the other not protein-coding. This gene has a pseudogene on chromosome 7.
Specifications
MTHFD2 | |
Polyclonal | |
Unconjugated | |
MTHFD2 | |
AW558851; Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase, mitochondrial; Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NAD+ dependent), methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase 2; Mthfd2; NAD-dependent methylene tetrahydrofolate dehydrogenase cyclohydrolase; NAD-dependent methylenetetrahydrofolate dehydrogenase; NMDMC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10797, 17768, 680308 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P13995, P18155 | |
MTHFD2 | |
Recombinant fusion protein of human MTHFD2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction