Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTHFD2L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544491
Description
Peptide sequence: TGIQTFGKNV VVAGRSKNVG MPIAMLLHTD GEHERPGGDA TVTIAHRYTP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
The specific function of this protein remains unknown.
Specifications
MTHFD2L | |
Polyclonal | |
Unconjugated | |
MTHFD2L | |
1110019K23Rik; C630010D07Rik; FLJ13105; LOW QUALITY PROTEIN: probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2 like; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; methylenetetrahydrofolate dehydrogenase 2-like; MGC45532; MGC72244; MTHFD2L; MTHFD2-like; NAD-dependent methylenetetrahydrofolate dehydrogenase; NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like; putative bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like protein; RGD1310879; tetrahydrofolate dehydrogenase/cyclohydrolase; Unknown (protein for MGC:63479); wu:fi14c12; zgc:63479 | |
Rabbit | |
Affinity chromatography | |
RUO | |
441024 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H903 | |
MTHFD2L | |
Synthetic peptide directed towards the middle region of human MTHFD2L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction