Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTHFD2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557269
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557269 Supplier Invitrogen™ Supplier No. PA557269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PASHTYVRNK IRAASAVGIC SELILKPKDV SQEELLDVTD QLNMDPRVSG ILVQLPLPDH VDERTI Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

The specific function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTHFD2L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTHFD2L
Gene Accession No. Q9H903
Gene Alias 1110019K23Rik; C630010D07Rik; FLJ13105; LOW QUALITY PROTEIN: probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2 like; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; methylenetetrahydrofolate dehydrogenase 2-like; MGC45532; MGC72244; MTHFD2L; MTHFD2-like; NAD-dependent methylenetetrahydrofolate dehydrogenase; NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like; putative bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like protein; RGD1310879; tetrahydrofolate dehydrogenase/cyclohydrolase; Unknown (protein for MGC:63479); wu:fi14c12; zgc:63479
Gene Symbols MTHFD2L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTHFD2L. Recombinant protein control fragment (Product #RP-96712).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441024
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.